2-Methyl-6-phenylpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2001-1G
M2001-5G
TCM2001-1GEA
216.46
CAD
TCM2001-1G
TCM2001-5G
2-Methyl-6-phenylpyridine ≥98.0% (by GC, titration analysis)
2-Methyl-6-phenylpyridine
CAS Number: 46181-30-0
MDL Number: MFCD00796537
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 155
Formula:
C₁₂H₁₁N Boiling Pt: 155 °C |
MDL Number:
MFCD00796537 CAS Number: 46181-30-0 EINECS: 256-258-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...