6-Methoxypurine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M1925-5G M1925-1G
TCM1925-5GEA 137.46 CAD
TCM1925-5G TCM1925-1G
6-Methoxypurine ≥98.0% (by HPLC, titration analysis)
6-Methoxypurine

CAS Number: 1074-89-1
MDL Number: MFCD00005574
Molecular Formula: C6H6N4O
Molecular Weight: 150.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 196

Formula: C₆H₆N₄O
Melting Pt: 196 °C
MDL Number: MFCD00005574
CAS Number: 1074-89-1
EINECS: 214-047-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f95836e28aa61b8ae89e7eba0714fdf5ce6e8c45&vid=f95836e28aa61b8ae89e7eba0714fdf5ce6e8c45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR