3-Methoxythioanisole ≥98.0%

Supplier: TCI America

M1880-25G M1880-5G
TCM1880-25GEA 137.46 CAD
TCM1880-25G TCM1880-5G
3-Methoxythioanisole ≥98.0%
3-Methoxythioanisole

CAS Number: 2388-74-1
MDL Number: MFCD03789162
Molecular Formula: C8H10OS
Molecular Weight: 154.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 3-(Methylthio)anisole

Formula: C₈H₁₀OS
Boiling Pt: 127 °C
Density: 1.12 (20 °C)
MDL Number: MFCD03789162
CAS Number: 2388-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32373e89bb3fe08293d3d546fc8c6c5c6462f56a&vid=32373e89bb3fe08293d3d546fc8c6c5c6462f56a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR