Methyl-2,4-difluorobenzoate ≥98.0%

Supplier: TCI America

M1879-5G M1879-25G
TCM1879-5GEA 69.52 CAD
TCM1879-5G TCM1879-25G
Methyl-2,4-difluorobenzoate ≥98.0%
Methyl 2,4-difluorobenzoate

CAS Number: 106614-28-2
MDL Number: MFCD03093799
Molecular Formula: C8H6F2O2
Molecular Weight: 172.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 82
Specific Gravity (20/20): 1.31


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4-Difluorobenzoic Acid Methyl Ester

Formula: C₈H₆F₂O₂
Boiling Pt: 194 °C
Density: 1.31 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD03093799
CAS Number: 106614-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac4fe93e034d7ef084d28a0951aeebc572cc038f&vid=ac4fe93e034d7ef084d28a0951aeebc572cc038f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR