7-Methoxyindole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 7-Methoxy-1H-indole, Indol-7-yl methyl ether

M1751-1G M1751-5G
TCM1751-1GEA 237 CAD
TCM1751-1G TCM1751-5G
7-Methoxyindole ≥98.0%
7-Methoxyindole

CAS Number: 3189-22-8
MDL Number: MFCD00047203
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Flash Point (°C): 113
Specific Gravity (20/20): 1.15

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 166 °C
Density: 1.15 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00047203
CAS Number: 3189-22-8
EINECS: 221-690-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8ecdf02b452a05069a9234ac6016f3ccc53d1b6&vid=b8ecdf02b452a05069a9234ac6016f3ccc53d1b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR