5-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M1676-25G M1676-5G
TCM1676-25GEA 129.56 CAD
TCM1676-25G TCM1676-5G
5-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Methylsalicylaldehyde

CAS Number: 613-84-3
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 218
Melting point (°C): 56


Other Product Names / Synonyms: 2-Hydroxy-5-methylbenzaldehyde

Formula: C₈H₈O₂
Boiling Pt: 218 °C
Melting Pt: 56 °C
Density: 1.0913 g/cm³ (59 °C)
Storage Temperature: Ambient
MDL Number: MFCD00274297
CAS Number: 613-84-3
EINECS: 210-357-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b667bbcc2fa18764d97c21cb2d56440ac0070aa&vid=17f7a6a91cbc33b32cd1bb4552286858803a7287&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR