5-Methoxy-2-tetralone ≥95.0%
Supplier: TCI America
M1543-1G
M1543-5G
TCM1543-1GEA
41.08
CAD
TCM1543-1G
TCM1543-5G
5-Methoxy-2-tetralone ≥95.0%
5-Methoxy-2-tetralone
CAS Number: 32940-15-1
MDL Number: MFCD00064958
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 37
Other Product Names / Synonyms: 3,4-Dihydro-5-methoxy-2(1H)-naphthalenone
Formula:
C₁₁H₁₂O₂ Boiling Pt: 165 °C Melting Pt: 37 °C |
MDL Number:
MFCD00064958 CAS Number: 32940-15-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...