2-Methylbenzo[b]thiophene ≥98.0%

Supplier: TCI America

M1429-1G M1429-5G
TCM1429-1GEA 79 CAD
TCM1429-1G TCM1429-5G
2-Methylbenzo[b]thiophene ≥98.0%
2-Methylbenzo[b]thiophene

CAS Number: 1195-14-8
MDL Number: MFCD00216250
Molecular Formula: C9H8S
Molecular Weight: 148.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Methylthianaphthene

Formula: C₉H₈S
Melting Pt: 55 °C
Flash Pt: 110 °C
MDL Number: MFCD00216250
CAS Number: 1195-14-8
EINECS: 214-792-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b869bf7c8a5d5ccba7909aa4d1f36ed6c173f4&vid=c2b869bf7c8a5d5ccba7909aa4d1f36ed6c173f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR