2-Methyl-1,3-propanediol ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dihydroxy-2-methylpropane

M1114-500ML M1114-25ML
TCM1114-500MLEA 72.21 CAD
TCM1114-500ML TCM1114-025ML
2-Methyl-1,3-propanediol ≥98.0%
2-Methyl-1,3-propanediol

CAS Number: 2163-42-0
MDL Number: MFCD00082586
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 110
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 1,3-Dihydroxy-2-methylpropane

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 214 °C
Density: 1.02 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00082586
CAS Number: 2163-42-0
EINECS: 412-350-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a84a8c7266f92c2bb211865a0ec5defb40d60817&vid=a84a8c7266f92c2bb211865a0ec5defb40d60817&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR