3-Methyl-2(5H)-furanone ≥97.0%

Supplier: TCI America

M1078-25ML M1078-5ML M1078-1ML
TCM1078-25MLEA 1341.42 CAD
TCM1078-25ML TCM1078-5ML TCM1078-001ML
3-Methyl-2(5H)-furanone ≥97.0%
3-Methyl-2(5H)-furanone

CAS Number: 22122-36-7
MDL Number: MFCD00191545
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 93
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 4-Hydroxy-2-methyl-2-butenoic Acid gamma-Lactone

Formula: C₅H₆O₂
Boiling Pt: 98 °C
Density: 1.12 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00191545
CAS Number: 22122-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a03ae7a709e0b72e0858f20c6bd3df83b6817dd0&vid=a03ae7a709e0b72e0858f20c6bd3df83b6817dd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR