2-Methoxy-1-naphthaldehyde ≥98.0%
Supplier: TCI America
M1040-5G
M1040-25G
TCM1040-005GEA
56.88
CAD
TCM1040-005G
TCM1040-025G
2-Methoxy-1-naphthaldehyde ≥98.0%
2-Methoxy-1-naphthaldehyde
CAS Number: 5392-12-1
MDL Number: MFCD00004004
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 84
Formula:
C₁₂H₁₀O₂ Boiling Pt: 205 °C Melting Pt: 84 °C |
MDL Number:
MFCD00004004 CAS Number: 5392-12-1 EINECS: 226-392-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...