Methyl-4-methoxyacetoacetate ≥97.0%
Supplier: TCI America
M1031-5G
M1031-25G
TCM1031-5GEA
72.68
CAD
TCM1031-5G
TCM1031-025G
Methyl-4-methoxyacetoacetate ≥97.0%
Methyl 4-methoxyacetoacetate
CAS Number: 41051-15-4
MDL Number: MFCD00010183
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 89
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: 4-Methoxyacetoacetic Acid Methyl Ester
Formula:
C₆H₁₀O₄ Boiling Pt: 113 °C Density: 1.13 (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00010183 CAS Number: 41051-15-4 EINECS: 255-188-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...