4-Methoxycinnamaldehyde ≥97.0%

Supplier: TCI America

Synonyms: 3-(4-Methoxyphenyl)-2-propenal, 3-(4-Methoxyphenyl)acrylaldehyde

M1012-25G M1012-5G
TCM1012-025GEA 202.24 CAD
TCM1012-025G TCM1012-5G
4-Methoxycinnamaldehyde ≥97.0%
4-Methoxycinnamaldehyde

CAS Number: 1963-36-6
MDL Number: MFCD00017343
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 176
Melting point (°C): 58
Flash Point (°C): 110


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(4-Methoxyphenyl)acrylaldehyde, 3-(4-Methoxyphenyl)-2-propenal

Formula: C₁₀H₁₀O₂
MW: 162.1852 g/mol
Boiling Pt: 176 °C
Melting Pt: 58 °C
Flash Pt: 110 °C
MDL Number: MFCD00017343
CAS Number: 1963-36-6
EINECS: 217-807-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e45f8c011f49afae684fcbc13345ad64509f4930&vid=e45f8c011f49afae684fcbc13345ad64509f4930&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR