Dimethyl methoxymalonate ≥95.0%
Supplier: TCI America
M0836-25G
M0836-5G
TCM0836-025GEA
697.89
CAD
TCM0836-025G
TCM0836-5G
Dimethyl methoxymalonate ≥95.0%
Dimethyl methoxymalonate
CAS Number: 5018-30-4
MDL Number: MFCD00009847
Molecular Formula: C6H10O5
Molecular Weight: 162.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 107
Freezing point (°C): 12
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: Methoxymalonic Acid Dimethyl Ester
Formula:
C₆H₁₀O₅ Boiling Pt: 110 °C Density: 1.18 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00009847 CAS Number: 5018-30-4 EINECS: 225-697-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...