3-Methyl-1,3-butanediol ≥98.0%
Supplier: TCI America
M0818-25ML
M0818-500ML
TCM0818-025MLEA
52.14
CAD
TCM0818-025ML
TCM0818-500ML
3-Methyl-1,3-butanediol ≥98.0%
3-Methyl-1,3-butanediol
CAS Number: 2568-33-4
MDL Number: MFCD00059655
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 203
Flash Point (°C): 116
Specific Gravity (20/20): 0.98
Formula:
C₅H₁₂O₂ Boiling Pt: 203 °C Density: 0.98 (20 °C) Flash Pt: 116 °C |
MDL Number:
MFCD00059655 CAS Number: 2568-33-4 EINECS: 459-270-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...