3-Methyl-1-phenyl-2-phospholene-1-oxide ≥95.0%

Supplier: TCI America

M0705-5G M0705-25G
TCM0705-005GEA 105.86 CAD
TCM0705-005G TCM0705-025G
3-Methyl-1-phenyl-2-phospholene-1-oxide ≥95.0%
3-Methyl-1-phenyl-2-phospholene 1-oxide

CAS Number: 707-61-9
MDL Number: MFCD00014518
Molecular Formula: C11H13OP
Molecular Weight: 192.20
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 64
Flash Point (°C): 110


Other Product Names / Synonyms: 2,3-Dihydro-4-methyl-1-phenyl-1H-phosphole 1-Oxide, 4-Methyl-1-phenyl-2,3-dihydrophosphol-1-one

Formula: C₁₁H₁₃OP
Boiling Pt: 155 °C
Melting Pt: 64 °C
Flash Pt: 110 °C
MDL Number: MFCD00014518
CAS Number: 707-61-9
EINECS: 211-901-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd1a3d7a8bbdbda8090b41d7b39d6abea2973428&vid=fd1a3d7a8bbdbda8090b41d7b39d6abea2973428&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR