3-Methoxycinnamic acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: 3-(3-Methoxyphenyl)acrylic acid

M0444-25G M0444-250G
TCM0444-025GEA 156.42 CAD
TCM0444-025G TCM0444-250G
3-Methoxycinnamic acid ≥99.0%
3-Methoxycinnamic acid

CAS Number: 6099-04-3
MDL Number: MFCD00004386
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 119


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: O-Methyl-m-coumaric Acid, O-Methyl-m-cumaric Acid

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00004386
CAS Number: 6099-04-3
EINECS: 228-049-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d19fbfdaafca1d9ec6bd877f0d250459f6f12bcb&vid=d19fbfdaafca1d9ec6bd877f0d250459f6f12bcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR