2-Methyl-1-nitronaphthalene ≥98.0%
Supplier: TCI America
M0376-250G
M0376-25G
TCM0376-250GEA
1094.31
CAD
TCM0376-250G
TCM0376-025G
2-Methyl-1-nitronaphthalene ≥98.0%
2-Methyl-1-nitronaphthalene
CAS Number: 881-03-8
MDL Number: MFCD00003914
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 188
Melting point (°C): 81
Formula:
C₁₁H₉NO₂ Boiling Pt: 188 °C Melting Pt: 81 °C |
MDL Number:
MFCD00003914 CAS Number: 881-03-8 EINECS: 212-917-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...