1-(Methylamino)anthraquinone ≥98.0%

Supplier: TCI America

Synonyms: 1-(Methylamino)anthracene-9,10-dione

M0255-500G M0255-25G
TCM0255-500GEA 667.71 CAD
TCM0255-500G TCM0255-025G
1-(Methylamino)anthraquinone ≥98.0%
1-(Methylamino)anthraquinone

CAS Number: 82-38-2
MDL Number: MFCD00001197
Molecular Formula: C15H11NO2
Molecular Weight: 237.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 169

Formula: C₁₅H₁₁NO₂
Melting Pt: 169 °C
MDL Number: MFCD00001197
CAS Number: 82-38-2
EINECS: 201-417-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0bd23909051c9c3575e1cc8003a27e97ee0e839&vid=d0bd23909051c9c3575e1cc8003a27e97ee0e839&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR