5-Methoxytryptamine

Supplier: TCI America

M0131-5G M0131-1G M0131-100MG
TCM0131-5GEA 292.3 CAD
TCM0131-5G TCM0131-001G TCM0131-100MG
5-Methoxytryptamine
5-Methoxytryptamine

CAS Number: 608-07-1
MDL Number: MFCD00005662
Molecular Formula: C11H14N2O
Molecular Weight: 190.25
Form: Crystal
Color: White
Melting point (°C): 121


Other Product Names / Synonyms: 3-(2-Aminoethyl)-5-methoxyindole, O-Methylserotonin

Formula: C₁₁H₁₄N₂O
MW: 190.24 g/mol
Melting Pt: 121 °C
MDL Number: MFCD00005662
CAS Number: 608-07-1
EINECS: 210-153-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4b13f4e6e1475dc8e9b93f7a2585be22d27271b&vid=e4b13f4e6e1475dc8e9b93f7a2585be22d27271b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR