Guaiacol ≥98.0%

Supplier: TCI America
Warning

M0121-500G M0121-25G
TCM0121-500GEA 86.9 CAD
TCM0121-500G TCM0121-025G
Guaiacol ≥98.0%
Guaiacol

CAS Number: 90-05-1
MDL Number: MFCD00002185
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 205
Flash Point (°C): 93
Freezing point (°C): 28


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Antioxidants, Others (Phytochemicals)
Other Product Names / Synonyms: Catechol Monomethyl Ether, 2-Hydroxyanisole, 2-Methoxyphenol, Pyrocatechol Monomethyl Ether

Formula: C₇H₈O₂
Boiling Pt: 205 °C
Melting Pt: 28…32 °C
Density: 1.1534 g/cm³ (0 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
UN: 2810
ADR: 6.1,III
Merck Index: 13,04568
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f516fcde314fb04777c33413bcc8c7af92fa090&vid=ff9544eee4966bddbc266b3dd9e068a2122cfaa3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR