2-Methoxynaphthalene ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-naphthyl ether

M0117-25G M0117-500G
TCM0117-025GEA 37.92 CAD
TCM0117-025G TCM0117-500G
2-Methoxynaphthalene ≥98.0%
2-Methoxynaphthalene

CAS Number: 93-04-9
MDL Number: MFCD00004061
Molecular Formula: C11H10O
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 274
Melting point (°C): 73


Other Product Names / Synonyms: Methyl 2-Naphthyl Ether

Formula: C₁₁H₁₀O
MW: 158.2 g/mol
Boiling Pt: 274 °C
Melting Pt: 73 °C
Density: 1.266 g/cm³ (20 °C)
MDL Number: MFCD00004061
CAS Number: 93-04-9
EINECS: 202-213-6
Merck Index: 12,06076
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ef33a75b64f15efe34e53e465678d524bcd2f80&vid=9ef33a75b64f15efe34e53e465678d524bcd2f80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR