Dibutyl maleate ≥95.0%

Supplier: TCI America

M0009-25G M0009-500G
TCM0009-025GEA 28.44 CAD
TCM0009-025G TCM0009-500G
Dibutyl maleate ≥95.0%
Dibutyl maleate

CAS Number: 105-76-0
MDL Number: MFCD00009447
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Melting point (°C): -85
Flash Point (°C): 140
Specific Gravity (20/20): 1.00


Product Categories: Materials Chemistry, Resin Additives, Copolymerization Monomers
Other Product Names / Synonyms: Maleic Acid Dibutyl Ester

Formula: C₁₂H₂₀O₄
Boiling Pt: 161 °C
Melting Pt: –85 °C
Density: 1.00 (20 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00009447
CAS Number: 105-76-0
EINECS: 203-328-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19b28a18b6f5c97e0d92db76931db83cdb02b8ce&vid=19b28a18b6f5c97e0d92db76931db83cdb02b8ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR