Butyl levulinate ≥98.0%

Supplier: TCI America

Synonyms: Levulinic acid butyl ester, 4-Oxovaleric acid butyl ester, Butyl 4-oxovalerate

L0138-25ML L0138-500ML
TCL0138-025MLEA 60.04 CAD
TCL0138-025ML TCL0138-500ML
Butyl levulinate ≥98.0%
Butyl levulinate

CAS Number: 2052-15-5
MDL Number: MFCD00009449
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Flash Point (°C): 120
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Levulinic Acid Butyl Ester, 4-Oxovaleric Acid Butyl Ester, Butyl 4-Oxovalerate

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
Boiling Pt: 237 °C
Density: 0.98 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00009449
CAS Number: 2052-15-5
EINECS: 218-143-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=108c1848fd1a78d73871b0ff44fa522268494d8d&vid=108c1848fd1a78d73871b0ff44fa522268494d8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR