Ethyl linoleate ≥97.0%

Supplier: TCI America

L0135-25ML L0135-5ML
TCL0135-025MLEA 559.51 CAD
TCL0135-025ML TCL0135-5ML
Ethyl linoleate ≥97.0%
Ethyl linoleate

CAS Number: 544-35-4
MDL Number: MFCD00009535
Molecular Formula: C20H36O2
Molecular Weight: 308.51
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 110
Specific Gravity (20/20): 0.88


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Linoleic Acid Ethyl Ester

Formula: C₂₀H₃₆O₂
Boiling Pt: 193 °C
Density: 0.88 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00009535
CAS Number: 544-35-4
EINECS: 208-868-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2806b2a57cc9bd2ebfc97c73fc661593a6ac495&vid=a2806b2a57cc9bd2ebfc97c73fc661593a6ac495&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR