3-Methyl-2-oxovaleric acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methyl-2-oxopentanoic acid

K0020-5G K0020-1G
TCK0020-005GEA 940.1 CAD
TCK0020-005G TCK0020-001G
3-Methyl-2-oxovaleric acid ≥95.0% (by GC, titration analysis)
3-Methyl-2-oxovaleric acid

CAS Number: 1460-34-0
MDL Number: MFCD00002582
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Boiling point (°C): 84
Melting point (°C): 42
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-Methyl-2-oxopentanoic Acid

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 84 °C
Melting Pt: 42 °C
MDL Number: MFCD00002582
CAS Number: 1460-34-0
EINECS: 215-955-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=664dd7219055249313031673e232faee2c264b37&vid=664dd7219055249313031673e232faee2c264b37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR