1-Iodoisoquinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
I0750-5G
I0750-25G
TCI0750-5GEA
370.19
CAD
TCI0750-5G
TCI0750-25G
1-Iodoisoquinoline ≥98.0% (by GC, titration analysis)
1-Iodoisoquinoline
CAS Number: 19658-77-6
Molecular Formula: C9H6IN
Molecular Weight: 255.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 75
Formula:
C₉H₆IN Melting Pt: 75 °C |
MDL Number:
MFCD00234494 CAS Number: 19658-77-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...