Indole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I0722-1G I0722-5G
TCI0722-1GEA 110.76 CAD
TCI0722-1G TCI0722-5G
Indole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Indole-4-carboxylic acid

CAS Number: 2124-55-2
MDL Number: MFCD00009739
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 212

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00009739
CAS Number: 2124-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f24d18788e0a7937c9d149bf5c86258792b74a1&vid=4f24d18788e0a7937c9d149bf5c86258792b74a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR