1-Isopropylnaphthalene ≥85.0%

Supplier: TCI America

I0578-5G I0578-1G
TCI0578-5GEA 436.08 CAD
TCI0578-5G TCI0578-1G
1-Isopropylnaphthalene ≥85.0%
1-Isopropylnaphthalene

CAS Number: 6158-45-8
MDL Number: MFCD00061059
Molecular Formula: C13H14
Molecular Weight: 170.26
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 268
Melting point (°C): -16
Specific Gravity (20/20): 0.99
Storage Temperature: 0-10°C

Formula: C₁₃H₁₄
MW: 170.25 g/mol
Boiling Pt: 268 °C
Melting Pt: –16 °C
Density: 0.99 (20 °C)
MDL Number: MFCD00061059
CAS Number: 6158-45-8
EINECS: 228-177-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7ca0f0df5a0b196db5a42a1c57fdd4ea41d71a1&vid=d7ca0f0df5a0b196db5a42a1c57fdd4ea41d71a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR