2-Iodohippuric acid ≥99.0%

Supplier: TCI America

Synonyms: N-(2-Iodobenzoyl)glycine

I0446-25G I0446-5G
TCI0446-25GEA 319.16 CAD
TCI0446-25G TCI0446-005G
2-Iodohippuric acid ≥99.0%
2-Iodohippuric acid

CAS Number: 147-58-0
MDL Number: MFCD00002694
Molecular Formula: C9H8INO3
Molecular Weight: 305.07
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 173


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-(2-Iodobenzoyl)glycine

Formula: C₉H₈INO₃
MW: 305.07 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00002694
CAS Number: 147-58-0
EINECS: 205-693-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19c0ebd1adb04d7775ffd3810ed5d12b1e461dd2&vid=19c0ebd1adb04d7775ffd3810ed5d12b1e461dd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR