Isovalerophenone ≥99.0%

Supplier: TCI America
Warning

I0296-25ML I0296-5ML
TCI0296-025MLEA 294.2 CAD
TCI0296-025ML TCI0296-5ML
Isovalerophenone ≥99.0%
Isovalerophenone

CAS Number: 582-62-7
MDL Number: MFCD00026486
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 236
Flash Point (°C): 99
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Isobutyl Phenyl Ketone

Formula: (C₆H₅)C(O)CH₂CH(CH₃)CH₃
Boiling Pt: 236 °C
Melting Pt: 2 °C
Density: 0.97 (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00026486
CAS Number: 582-62-7
EINECS: 209-489-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=428e5d56e1d402fb97e9df784edb6cf4195fb3e5&vid=428e5d56e1d402fb97e9df784edb6cf4195fb3e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR