4-Isopropylpyridine ≥98.0%

Supplier: TCI America
Warning

I0174-5G I0174-25G
TCI0174-5GEA 472.74 CAD
TCI0174-5G TCI0174-025G
4-Isopropylpyridine ≥98.0%
4-Isopropylpyridine

CAS Number: 696-30-0
MDL Number: MFCD00039719
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 66
Specific Gravity (20/20): 0.93

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 178 °C
Density: 0.93 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00039719
CAS Number: 696-30-0
EINECS: 211-794-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90640699e5ae612ece7268e642f04815181eee8a&vid=90640699e5ae612ece7268e642f04815181eee8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR