Methyl isonicotinate ≥99.0%

Supplier: TCI America

Synonyms: 4-Pyridinecarboxylic acid methyl ester, 4-Picolinic acid methyl ester, Methyl 4-pyridinecarboxylate, Pyridine-4-carboxylic acid methyl ester, Isonicotinic acid methyl ester

I0141-25G I0141-100G
TCI0141-025GEA 53.72 CAD
TCI0141-025G TCI0141-100G
Methyl isonicotinate ≥99.0%
Methyl isonicotinate

CAS Number: 2459-09-8
MDL Number: MFCD00006427
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Boiling point (°C): 209
Flash Point (°C): 82
Freezing point (°C): 15
Specific Gravity (20/20): 1.16
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isonicotinic Acid Methyl Ester, Methyl Pyridine-4-carboxylate, Pyridine-4-carboxylic Acid Methyl Ester

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 207
Melting Pt: 17
Density: 1.16 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00006427
CAS Number: 2459-09-8
EINECS: 219-546-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e33ef6385f479d7f47b789887ec5b789cc160c2&vid=7e33ef6385f479d7f47b789887ec5b789cc160c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR