1-Iodooctadecane ≥98.0%

Supplier: TCI America
Warning

Synonyms: Stearyl Iodide, Octadecyl Iodide

I0065-25G I0065-100G
TCI0065-025GEA 227.52 CAD
TCI0065-025G TCI0065-500G TCI0065-100G
1-Iodooctadecane ≥98.0%
1-Iodooctadecane

CAS Number: 629-93-6
MDL Number: MFCD00001090
Molecular Formula: C18H37I
Molecular Weight: 380.40
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 197
Melting point (°C): 34
Flash Point (°C): 110


Other Product Names / Synonyms: Octadecyl Iodide, Stearyl Iodide

Formula: C₁₈H₃₇I
MW: 380.4 g/mol
Boiling Pt: 197 °C
Melting Pt: 34 °C
Density: 1.104 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001090
CAS Number: 629-93-6
EINECS: 211-117-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc18c2e8840a84e14dd8919c6514911e5b4585ab&vid=cc18c2e8840a84e14dd8919c6514911e5b4585ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR