2-Imidazolidinone ≥97.0%

Supplier: TCI America

Synonyms: 2-Imidazolidone

I0005-25G I0005-500G
TCI0005-025GEA 34.76 CAD
TCI0005-025G TCI0005-500G
2-Imidazolidinone ≥97.0%
2-Imidazolidinone

CAS Number: 120-93-4
MDL Number: MFCD00005257
Molecular Formula: C3H6N2O
Molecular Weight: 86.09
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 132
Flash Point (°C): 265


Other Product Names / Synonyms: Ethyleneurea, 2-Imidazolidone

Formula: C₃H₆N₂O
MW: 86.0925 g/mol
Melting Pt: 132 °C
Flash Pt: 265 °C
MDL Number: MFCD00005257
CAS Number: 120-93-4
EINECS: 204-436-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0265860465ae5fad25d1c5811b6783447a026307&vid=0265860465ae5fad25d1c5811b6783447a026307&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR