1-Hexyl-3-methylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
H1227-25G
H1227-5G
TCH1227-25GEA
619.83
CAD
TCH1227-25G
TCH1227-5G
1-Hexyl-3-methylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
1-Hexyl-3-methylimidazolium bromide
CAS Number: 85100-78-3
MDL Number: MFCD03427612
Molecular Formula: C10H19BrN2
Molecular Weight: 247.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Boiling point (°C): 395
Formula:
C₁₀H₁₉BrN₂ Boiling Pt: 395 °C |
MDL Number:
MFCD03427612 CAS Number: 85100-78-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...