4'-Hydroxy-2'-methylacetophenone ≥98.0%
Supplier: TCI America
Synonyms:
4′-Hydroxy-2′-methylacetophenone, 1-(4-Hydroxy-2-methylphenyl)ethanone
H1124-25G
H1124-5G
TCH1124-25GEA
464.52
CAD
TCH1124-25G
TCH1124-5G
4'-Hydroxy-2'-methylacetophenone ≥98.0%
4'-Hydroxy-2'-methylacetophenone
CAS Number: 875-59-2
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 313
Melting point (°C): 130
Other Product Names / Synonyms: 4-Acetyl-3-methylphenol
Formula:
C₉H₁₀O₂ Boiling Pt: 313 °C Melting Pt: 130 °C |
MDL Number:
MFCD00002303 CAS Number: 875-59-2 EINECS: 212-874-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...