4-Hydroxycarbazole ≥98.0% (by HPLC)

Supplier: TCI America

H1014-1G H1014-25G H1014-5G
TCH1014-1GEA 33.18 CAD
TCH1014-1G TCH1014-25G TCH1014-5G
4-Hydroxycarbazole ≥98.0% (by HPLC)
4-Hydroxycarbazole

CAS Number: 52602-39-8
MDL Number: MFCD02178385
Molecular Formula: C12H9NO
Molecular Weight: 183.21
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 169


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 9H-Carbazol-4-ol

Formula: C₁₂H₉NO
Melting Pt: 169 °C
MDL Number: MFCD02178385
CAS Number: 52602-39-8
EINECS: 258-034-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105678ae14c4b258549f801ccd2737feccfdb316&vid=105678ae14c4b258549f801ccd2737feccfdb316&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR