(S,S)-(-)-Hydrobenzoin ≥99.0%

Supplier: TCI America

H0824-25G H0824-5G
TCH0824-25GEA 960.64 CAD
TCH0824-25G TCH0824-5G
(S,S)-(-)-Hydrobenzoin ≥99.0%
(S,S)-(-)-Hydrobenzoin

CAS Number: 2325-10-2
MDL Number: MFCD00064255
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 148
Specific rotation [a]20/D: -95 deg (C=1, CHCl3)


Other Product Names / Synonyms: (S,S)-(-)-1,2-Diphenyl-1,2-ethanediol

Formula: C₁₄H₁₄O₂
Melting Pt: 148 °C
MDL Number: MFCD00064255
CAS Number: 2325-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1326530aa2eb3b366ddc64f2685da3f40156aa1&vid=f1326530aa2eb3b366ddc64f2685da3f40156aa1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR