(R,R)-(+)-Hydrobenzoin ≥99.0%
Supplier: TCI America
Synonyms:
(1R,2R)-1,2-Diphenylethane-1,2-diol, (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol
H0818-25G
H0818-5G
TCH0818-25GEA
423.44
CAD
TCH0818-25G
TCH0818-5G
(R,R)-(+)-Hydrobenzoin ≥99.0%
(R,R)-(+)-Hydrobenzoin
CAS Number: 52340-78-0
MDL Number: MFCD00064254
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 148
Specific rotation [a]20/D: 95 deg (C=2, EtOH)
Other Product Names / Synonyms: (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol
Formula:
C₁₄H₁₄O₂ Melting Pt: 148 °C |
MDL Number:
MFCD00064254 CAS Number: 52340-78-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...