4-Hydroxyphthalonitrile ≥97.0%
Supplier: TCI America
|
Warning
|
H0813-5G
H0813-1G
TCH0813-005GEA
423.44
CAD
TCH0813-005G
TCH0813-001G
4-Hydroxyphthalonitrile ≥97.0%
4-Hydroxyphthalonitrile
CAS Number: 30757-50-7
MDL Number: MFCD00191653
Molecular Formula: C8H4N2O
Molecular Weight: 144.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 212
Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 3,4-Dicyanophenol
Formula:
C₈H₄N₂O Melting Pt: 212 °C |
MDL Number:
MFCD00191653 CAS Number: 30757-50-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...