4'-Hydroxyvalerophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(4-Hydroxyphenyl)-1-pentanone

H0627-500G H0627-25G
TCH0627-500GEA 1726.94 CAD
TCH0627-500G TCH0627-025G
4'-Hydroxyvalerophenone ≥98.0%
4'-Hydroxyvalerophenone

CAS Number: 2589-71-1
MDL Number: MFCD00009719
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 66


Other Product Names / Synonyms: 4'-Hydroxypentanophenone

Formula: C₁₁H₁₄O₂
Boiling Pt: 156…158 °C (0.01 mmHg)
Melting Pt: 66 °C
MDL Number: MFCD00009719
CAS Number: 2589-71-1
EINECS: 219-978-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9608ae71fb1a19d07f4c18777529a8a2e4cfb8bf&vid=9608ae71fb1a19d07f4c18777529a8a2e4cfb8bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR