Methyl heptadecanoate ≥97.0%

Supplier: TCI America

H0566-5G H0566-25G
TCH0566-005GEA 136.16 CAD
TCH0566-005G TCH0566-025G
Methyl heptadecanoate ≥97.0%
Methyl heptadecanoate

CAS Number: 1731-92-6
MDL Number: MFCD00009001
Molecular Formula: C18H36O2
Molecular Weight: 284.48
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 28


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Heptadecanoic Acid Methyl Ester, Margaric Acid Methyl Ester, Methyl Margarate

Formula: C₁₈H₃₆O₂
Flash Pt: 110 °C
MDL Number: MFCD00009001
CAS Number: 1731-92-6
EINECS: 217-055-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a35d1518b271bd97bc4b36f1775b2ca8ca0dc200&vid=a35d1518b271bd97bc4b36f1775b2ca8ca0dc200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR