1,16-Hexadecanediol ≥95.0%

Supplier: TCI America

H0552-1G H0552-5G
TCH0552-001GEA 172.22 CAD
TCH0552-001G TCH0552-5G
1,16-Hexadecanediol ≥95.0%
1,16-Hexadecanediol

CAS Number: 7735-42-4
MDL Number: MFCD00002821
Molecular Formula: C16H34O2
Molecular Weight: 258.45
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 92


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,16-Dihydroxyhexadecane, Hexadecamethylene Glycol

Formula: C₁₆H₃₄O₂
Boiling Pt: 200 °C
Melting Pt: 92 °C
MDL Number: MFCD00002821
CAS Number: 7735-42-4
EINECS: 231-794-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=638b4b367e629c89d37ffd9e8ad928895eb2fc79&vid=638b4b367e629c89d37ffd9e8ad928895eb2fc79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR