11-Heneicosanone ≥95.0%

Supplier: TCI America

Synonyms: Didecyl ketone

H0532-5G H0532-25G
TCH0532-5GEA 366.56 CAD
TCH0532-5G TCH0532-025G
11-Heneicosanone ≥95.0%
11-Heneicosanone

CAS Number: 19781-72-7
MDL Number: MFCD00048410
Molecular Formula: C21H42O
Molecular Weight: 310.57
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 64


Other Product Names / Synonyms: Didecyl Ketone

Formula: C₂₁H₄₂O
Melting Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00048410
CAS Number: 19781-72-7
EINECS: 243-307-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4d66bbf0a9ec019f5650e6889728319110d5848&vid=888533d8d232f5130eee7a17101451b9bb326873&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR