3-Hexyn-1-ol ≥97.0%

Supplier: TCI America
Warning

H0460-25ML H0460-5ML
TCH0460-025MLEA 1899.16 CAD
TCH0460-025ML TCH0460-5ML
3-Hexyn-1-ol ≥97.0%
3-Hexyn-1-ol

CAS Number: 1002-28-4
MDL Number: MFCD00002957
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 64
Specific Gravity (20/20): 0.90

Formula: C₆H₁₀O
Boiling Pt: 161 °C
Melting Pt: 42 °C
Density: 0.90 (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00002957
CAS Number: 1002-28-4
EINECS: 213-685-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e3e87241988608641a7f88b3b3bf2bbe44a890&vid=d4e3e87241988608641a7f88b3b3bf2bbe44a890&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR