2,2,4,4,6,8,8-Heptamethylnonane ≥97.0%

Supplier: TCI America
Danger

H0365-25ML H0365-500ML H0365-100ML
TCH0365-025MLEA 75.84 CAD
TCH0365-025ML TCH0365-500ML 76220-108
2,2,4,4,6,8,8-Heptamethylnonane ≥97.0%
2,2,4,4,6,8,8-Heptamethylnonane

CAS Number: 4390-04-9
MDL Number: MFCD00008856
Molecular Formula: C16H34
Molecular Weight: 226.45
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 247
Flash Point (°C): 102
Specific Gravity (20/20): 0.78

Formula: C₁₆H₃₄
Boiling Pt: 247 °C
Density: 0.78 (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00008856
CAS Number: 4390-04-9
EINECS: 224-506-8
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b09c00f3b2d72a79b71ea23bd4af3d7c7305a82&vid=290f3760c5c81385fa5967e31a5843c8af43be8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR