1-Hexacosanol ≥95.0%
Supplier: TCI America
H0342-100MG
H0342-1G
TCH0342-100MGEA
275.06
CAD
TCH0342-100MG
TCH0342-001G
1-Hexacosanol ≥95.0%
1-Hexacosanol
CAS Number: 506-52-5
MDL Number: MFCD00002940
Molecular Formula: C26H54O
Molecular Weight: 382.72
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 80
Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Ceryl Alcohol
Formula:
H₃C(CH₂)₂₄CH₂OH Boiling Pt: 412 °C (1013 hPa) Melting Pt: 80 °C Density: 0.8215 g/cm³ (85 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002940 CAS Number: 506-52-5 EINECS: 208-044-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...