2-Hydroxy-1-naphthoic acid ≥98.0%

Supplier: TCI America

H0280-250G H0280-25G
TCH0280-250GEA 308.1 CAD
TCH0280-250G TCH0280-025G
2-Hydroxy-1-naphthoic acid ≥98.0%
2-Hydroxy-1-naphthoic acid

CAS Number: 2283-08-1
MDL Number: MFCD00004009
Molecular Formula: C11H8O3
Molecular Weight: 188.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 167


Other Product Names / Synonyms: 2-Hydroxy-1-naphthalenecarboxylic Acid

Formula: C₁₁H₈O₃
Melting Pt: 167 °C
MDL Number: MFCD00004009
CAS Number: 2283-08-1
EINECS: 218-919-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=269d06ad0ea6610243b25a86e5bfffdb00507a22&vid=269d06ad0ea6610243b25a86e5bfffdb00507a22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR