Hexyl-4-hydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxybenzoic acid hexyl ester, Hexyl 4-hydroxybenzoate, Hexylparaben

H0213-500G H0213-25G H0213-100G
TCH0213-500GEA 752.08 CAD
TCH0213-500G TCH0213-025G TCH0213-100G
Hexyl-4-hydroxybenzoate ≥98.0%
Hexyl 4-hydroxybenzoate

CAS Number: 1083-27-8
MDL Number: MFCD00016480
Molecular Formula: C13H18O3
Molecular Weight: 222.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 171
Melting point (°C): 53


Other Product Names / Synonyms: Hexylparaben, 4-Hydroxybenzoic Acid Hexyl Ester

Formula: C₁₃H₁₈O₃
Boiling Pt: 171 °C
Melting Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00016480
CAS Number: 1083-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ef77c88501c20fbe7ce12f7e99119fea7186635&vid=6ef77c88501c20fbe7ce12f7e99119fea7186635&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR